Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SULT1A1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572202
Description
The human Catecholamine-Sulfating Phenol Sulfotransferase (CSPS) is the only sulfotransferase that catalyzes the sulfation of catecholamins, in particular the neurotransmitter dopamine, with high activity. CSPS is required for stimulation by Mn2+ of the sulfating activity and expressed in the human intestine, brain, platelet and other tissues. In the brain it may play a role in regulating the levels of dopamine. It also serves as a detoxifying function in the intestine, where it may detoxify potentially lethal dietary monoamines.
Specifications
SULT1A1 | |
Polyclonal | |
Unconjugated | |
SULT1A1 | |
Sulfotransferase 1A1; ST1A1; Aryl sulfotransferase 1; HAST1/HAST2; Phenol sulfotransferase 1; Phenol-sulfating phenol sulfotransferase 1; P-PST 1; ST1A3; Thermostable phenol sulfotransferase; Ts-PST; SULT1A1; STP; STP1 | |
Rabbit | |
Protein A | |
RUO | |
6817 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Western Blot | |
0.92 mg/mL | |
PBS with 0.09% sodium azide | |
P50225 | |
SULT1A1 | |
KLH conjugated synthetic peptide between 246-279 amino acids from the C-terminal region of human SULT1A1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction