Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SULT1A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588806
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588806 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588806 Supplier Invitrogen™ Supplier No. PA588806
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KREIQKILEF VGRSLPEETV DLMVEHTSFK EMKKNPMTNY TTVRREFMDH SISPFMRKGM AGDWKTTFTV AQNERFDADY AEKMAGCSLS FRSEL; Positive Samples: A-431, Mouse liver; Cellular Location: Cytoplasm.

Sulfotransferase that utilizes 3'-phospho-5'-adenylyl sulfate (PAPS) as sulfonate donor to catalyze the sulfate conjugation of catecholamines, phenolic drugs and neurotransmitters. Is also responsible for the sulfonation and activation of minoxidil. Mediates the metabolic activation of carcinogenic N-hydroxyarylamines to DNA binding products and could so participate as modulating factor of cancer risk.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SULT1A2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.97 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SULT1A2
Gene Accession No. P50226
Gene Alias aryl sulfotransferase 2; arylamine sulfotransferase; HAST4; phenol sulfotransferase 2; phenolic-metabolizing (P) form of PST; phenol-preferring phenol sulfotransferase2; phenol-sulfating phenol sulfotransferase 2; P-PST; P-PST 2; ST1A2; STP2; sulfotransferase 1A2; sulfotransferase family 1A member 2; sulfotransferase family, cytosolic, 1A, phenol-preferring, member 2; SULT1A2; thermostable phenol sulfotransferase; TSPST2
Gene Symbols SULT1A2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6799
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.