Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SULT1A2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2836040.1ML
Description
SULT1A2 Polyclonal specifically detects SULT1A2 in Human samples. It is validated for Western Blot.Specifications
SULT1A2 | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
6799 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Aryl sulfotransferase 2, arylamine sulfotransferase, EC 2.8.2, EC 2.8.2.1, HAST4, MGC142287, MGC142289, Phenol sulfotransferase 2, phenolic-metabolizing (P) form of PST, phenol-preferring phenol sulfotransferase2, Phenol-sulfating phenol sulfotransferase 2, P-PST 2, ST1A2, STP2P-PST, sulfotransferase 1A2, sulfotransferase family, cytosolic, 1A, phenol-preferring, member 2, thermostable phenol sulfotransferase, TSPST2 | |
The immunogen is a synthetic peptide directed towards the middle region of Human SULT1A2 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction