Learn More
Invitrogen™ SULT1A3 Monoclonal Antibody (OTI3G5)

Description
SULT1A3 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | SULT1A3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI3G5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SULT1A3 |
| Gene Accession No. | P0DMM9 |
| Gene Alias | aryl sulfotransferase 1A3/1A4; Catecholamine-sulfating phenol sulfotransferase; CSPS; dopamine-specific sulfotransferase; HAST; HAST3; Monoamine-sulfating phenol sulfotransferase; monoamine-sulfating phenosulfotransferase; M-PST; phenol sulfotransferase 1A5; placental estrogen sulfotransferase; ST1A3; ST1A3/ST1A4; ST1A4; ST1A5; STM; sulfokinase; sulfotransferase 1A3; Sulfotransferase 1A3/1A4; Sulfotransferase 1A4; sulfotransferase family 1A member 3; sulfotransferase family, cytosolic, 1A, phenol-preferring, member 3; Sulfotransferase, monoamine-preferring; SULT1A3; SULT1A4; thermolabile (monoamine, M form) phenol sulfotransferase; Thermolabile phenol sulfotransferase; TL-PST |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.