Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SULT1A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527479
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527479 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527479 Supplier Invitrogen™ Supplier No. PA527479
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HepG2. Predicted reactivity: Pig (81%), Rhesus Monkey (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene encodes a phenol sulfotransferase with thermolabile enzyme activity. Four sulfotransferase genes are located on the p arm of chromosome 16; this gene and SULT1A4 arose from a segmental duplication. This gene is the most centromeric of the four sulfotransferase genes. Exons of this gene overlap with exons of a gene that encodes a protein containing GIY-YIG domains (GIYD1). Multiple alternatively spliced variants that encode the same protein have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SULT1A3
Applications Western Blot
Classification Polyclonal
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene SULT1A3
Gene Accession No. P0DMM9
Gene Alias aryl sulfotransferase 1A3/1A4; Catecholamine-sulfating phenol sulfotransferase; CSPS; dopamine-specific sulfotransferase; HAST; HAST3; Monoamine-sulfating phenol sulfotransferase; monoamine-sulfating phenosulfotransferase; M-PST; phenol sulfotransferase 1A5; placental estrogen sulfotransferase; ST1A3; ST1A3/ST1A4; ST1A4; ST1A5; STM; sulfokinase; sulfotransferase 1A3; Sulfotransferase 1A3/1A4; Sulfotransferase 1A4; sulfotransferase family 1A member 3; sulfotransferase family, cytosolic, 1A, phenol-preferring, member 3; Sulfotransferase, monoamine-preferring; SULT1A3; SULT1A4; thermolabile (monoamine, M form) phenol sulfotransferase; Thermolabile phenol sulfotransferase; TL-PST
Gene Symbols SULT1A3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SULT1A3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6818
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.