Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SULT1B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547796
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547796 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547796 Supplier Invitrogen™ Supplier No. PA547796
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) SULT1A1 and rhSULT1A3 is observed and less than 2% cross-reactivity with rhSULT2A1, rhSULT4A1, rhSULT2B1, and rhSULT1E1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure is similar among family members. However, the total genomic length of this gene is greater than that of other SULT1 genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SULT1B1
Applications Neutralization, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene SULT1B1
Gene Accession No. O43704
Gene Alias DOPA/tyrosine sulfotransferase; ST1B1; ST1B2; St2b2; sulfotransferase 1B1; sulfotransferase 1B2; sulfotransferase family 1B member 1; sulfotransferase family 1B, member 1; sulfotransferase family cytosolic 1B member 1; sulfotransferase family, cytosolic, 1B, member 1; SULT1B1; SULT1B2; Thyroid hormone sulfotransferase
Gene Symbols SULT1B1
Host Species Sheep
Immunogen E. coli-derived recombinant human Cytosolic Sulfotransferase 1B1/SULT1B1 (aa 2-296).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27284
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.