Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SULT1B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113876
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113876 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113876 Supplier Invitrogen™ Supplier No. PA5113876
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLSPKDILRK DLKLVHGYPM TCAFASNWEK IEQFHSRPDD IVIATYPKSG For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 92%; Dog: 86%; Human: 100%; Pig: 79%; Rat: 86%.

Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure is similar among family members. However, the total genomic length of this gene is greater than that of other SULT1 genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SULT1B1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SULT1B1
Gene Accession No. O43704
Gene Alias DOPA/tyrosine sulfotransferase; ST1B1; ST1B2; St2b2; sulfotransferase 1B1; sulfotransferase 1B2; sulfotransferase family 1B member 1; sulfotransferase family 1B, member 1; sulfotransferase family cytosolic 1B member 1; sulfotransferase family, cytosolic, 1B, member 1; SULT1B1; SULT1B2; Thyroid hormone sulfotransferase
Gene Symbols SULT1B1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human SULT1B1 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27284
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.