Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SULT1C2 Monoclonal Antibody (OTI4H4), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene TA503094

Encompass_Preferred

 View more versions of this product

Catalog No. 50-167-4063


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene encodes a protein that belongs to the SULT1 subfamily, responsible for transferring a sulfo moiety from PAPS to phenol-containing compounds.
Specifications

Specifications

SULT1C2
Monoclonal
0.41 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O00338
SULT1C2
Full length human recombit protein of human SULT1C2 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2b
Flow Cytometry, Western Blot
OTI4H4
Unconjugated
SULT1C2
ST1C1, ST1C2, SULT1C1, humSULTC2
Mouse
Affinity Chromatography
RUO
6819
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.