Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SULT1C2 Monoclonal Antibody (OTI4H4), TrueMAB™, OriGene
SDP

Catalog No. 502541123
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1123 30 μL
50-167-4063 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1123 Supplier OriGene Supplier No. TA503094S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SULT1C2 Monoclonal antibody specifically detects SULT1C2 in Human, Mouse samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen SULT1C2
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI4H4
Concentration 0.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TAGLN
Gene Accession No. P37804, Q01995
Gene Alias (PST)G; 1810008N17Rik; 1C member 4; HAST-I; humSULTC2; LOC100729651; MGC149521; MGC34422; mOLFST; N-hydroxyarylamine sulfotransferase; N-hydroxylarylamine sulfotransferase; phenol sulfotransferase; Phenol sulfotransferase 1c1; P-SULT; rabSULT1C2; rSULT1C2; St1c1; ST1C2; ST1C4; Stp2; sulfotransferase; sulfotransferase 1C1; Sulfotransferase 1C2; sulfotransferase 1C4; sulfotransferase family 1A phenol-preferring member 2; sulfotransferase family 1A, member 2; sulfotransferase family 1A, phenol-preferring, member 2; sulfotransferase family 1C member 2; sulfotransferase family 1C member 4; sulfotransferase family, cytosolic, 1C, member 1; sulfotransferase family, cytosolic, 1C, member 2; sulfotransferase family, cytosolic, 1C, member 3; sulfotransferase family, cytosolic, 1C, member 4; sulfotransferase family, cytosolic, 1C, member C2; sulfotransferase K1; sulfotransferase, phenol preferring 2; sulphotransferase family cytosolic 1A phenol-preferring member 2; Sult1a2; SULT1C; SULT1C#1; SULT1C#2; SULT1C1; SULT1C2; Sult1c3; SULT1C4; Sultk1
Host Species Mouse
Immunogen Full length human recombinant protein of human TAGLN produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21345, 6876
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.