Learn More
Invitrogen™ SULT1C2 Monoclonal Antibody (OTI5A4)

Description
SULT1C2 Monoclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | SULT1C2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI5A4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SULT1C2 |
| Gene Accession No. | O00338 |
| Gene Alias | (PST)G; 1810008N17Rik; 1C member 4; HAST-I; humSULTC2; LOC100729651; MGC149521; MGC34422; mOLFST; N-hydroxyarylamine sulfotransferase; N-hydroxylarylamine sulfotransferase; phenol sulfotransferase; Phenol sulfotransferase 1c1; P-SULT; rabSULT1C2; rSULT1C2; St1c1; ST1C2; ST1C4; Stp2; sulfotransferase; sulfotransferase 1C1; Sulfotransferase 1C2; sulfotransferase 1C4; sulfotransferase family 1A phenol-preferring member 2; sulfotransferase family 1A, member 2; sulfotransferase family 1A, phenol-preferring, member 2; sulfotransferase family 1C member 2; sulfotransferase family 1C member 4; sulfotransferase family, cytosolic, 1C, member 1; sulfotransferase family, cytosolic, 1C, member 2; sulfotransferase family, cytosolic, 1C, member 3; sulfotransferase family, cytosolic, 1C, member 4; sulfotransferase family, cytosolic, 1C, member C2; sulfotransferase K1; sulfotransferase, phenol preferring 2; sulphotransferase family cytosolic 1A phenol-preferring member 2; Sult1a2; SULT1C; SULT1C#1; SULT1C#2; SULT1C1; SULT1C2; Sult1c3; SULT1C4; Sultk1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.