Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SULT1E1 Recombinant Rabbit Monoclonal Antibody (022), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA543144
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543144 100 μL
PIMA543423 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543144 Supplier Invitrogen™ Supplier No. MA543144
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Specificity: Human SULT1E1

Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene encodes a protein that transfers a sulfo moiety to and from estrone, which may control levels of estrogen receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SULT1E1
Applications ELISA
Classification Recombinant Monoclonal
Clone 022
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Sult1e1
Gene Accession No. P49888
Gene Alias EST; EST-1; EST-2; EST-3; EST-6; estrogen sulfotransferase; Estrogen sulfotransferase, isoform 3; estrogen sulfotransferase, testis isoform; estrone sulfotransferase; ST1E1; Ste; ste2; Sulfotransferase 1E1; sulfotransferase family 1E member 1; sulfotransferase family 1E, estrogen-preferring, member 1; sulfotransferase family 1E, member 1; sulfotransferase, estrogen preferring; sulfotransferase, estrogen-preferring; SULT1E1
Gene Symbols Sult1e1
Host Species Rabbit
Immunogen Recombinant Human SULT1E1 Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6783
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.