Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SULT2A1 Monoclonal Antibody (9G8G12G10)
GREENER_CHOICE

Catalog No. PIMA529591
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529591 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529591 Supplier Invitrogen™ Supplier No. MA529591
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human SULT2A1/STD.

SULT1 sulfotransferases exhibit N-sulfating activities of carcinogenic heterocyclic amines, and are selective toward phenols, whereas SULT2 enzymes prefer hydroxysteroids and SULT3 family members are selective for N-substituted aryl and alicyclic compounds. SULT2A1 catalyzes the sulfonation of procarcinogen xenobiotics, hydroxysteroids and bile acids, and is highly expressed in adrenal and liver tissues. SULT2A1 plays a role in hepatic cholesterol homeostasis. SULT2B1 consists of two isoforms, SULT2B1a and SULT2B1b, which are transcribed from the same gene by alternative splicing of their first exons. Both isoforms are highly selective for the sulphation of 3b-hydroxysteroids such as pregnenolone, epiandrosterone, DHEA and androstenediol. SULT2B1b is expressed in prostate, skin, placenta and lung.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SULT2A1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 9G8G12G10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene SULT2A1
Gene Accession No. Q06520
Gene Alias alcohol sulfotransferase; alcohol/hydroxysteroid sulfotransferase; bile salt sulfotransferase; Bile salt sulfotransferase 1; bile-salt sulfotranasferase 2A1; bile-salt sulfotransferase 2A1; Dehydroepiandrosterone sulfotransferase; DHEAS; DHEA-ST; HST; hSTa; Hydroxysteroid sulfotransferase; hydroxysteroid sulfotransferase a; mSTa1; Smp-2; ST; ST2; ST-20; ST2A1; ST2A3; STa; Sta1; Std; Sth1; Sth2; Sulfotransferase 2A1; sulfotransferase family 2A member 1; sulfotransferase family 2A, dehydroepiandrosterone (DHEA)-preferring, member 1; sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone (DHEA) -preferring, member 1; sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone (DHEA)-preferring, member 1; sulfotransferase hydroxysteroid gene 2; sulfotransferase, DHEA preferring; sulfotransferase, hydroxysteroid preferring 1; sulfotransferase, hydroxysteroid preferring 2; SULT2A1
Gene Symbols SULT2A1
Host Species Mouse
Immunogen Recombinant Human SULT2A1 protein (Ser2-Glu285).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6822
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.