Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SULT2A1 Monoclonal Antibody (OTI3D4), TrueMAB™, OriGene
SDP

Catalog No. 501672833
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-2833 100 μL
50-254-2075 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-2833 Supplier OriGene Supplier No. TA501714
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene is primarily expressed in liver and adrenal tissues where the encoded protein sulfates steroids and bile acids.

Specifications

Antigen SULT2A1
Applications Western Blot
Classification Monoclonal
Clone OTI3D4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SULT2A1
Gene Accession No. Q06520
Gene Alias DHEA-ST, DHEAS, HST, ST2, ST2A1, ST2A3, STD, hSTa
Gene Symbols SULT2A1
Host Species Mouse
Immunogen Full length human recombit protein of human SULT2A1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6822
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.