Learn More
SULT2A1 Monoclonal Antibody (OTI3G4)

Description
SULT2A1 Monoclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | SULT2A1 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | OTI3G4 |
| Concentration | 0.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SULT2A1 |
| Gene Accession No. | Q06520 |
| Gene Alias | alcohol sulfotransferase; alcohol/hydroxysteroid sulfotransferase; bile salt sulfotransferase; Bile salt sulfotransferase 1; bile-salt sulfotranasferase 2A1; bile-salt sulfotransferase 2A1; Dehydroepiandrosterone sulfotransferase; DHEAS; DHEA-ST; HST; hSTa; Hydroxysteroid sulfotransferase; hydroxysteroid sulfotransferase a; mSTa1; Smp-2; ST; ST2; ST-20; ST2A1; ST2A3; STa; Sta1; Std; Sth1; Sth2; Sulfotransferase 2A1; sulfotransferase family 2A member 1; sulfotransferase family 2A, dehydroepiandrosterone (DHEA)-preferring, member 1; sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone (DHEA) -preferring, member 1; sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone (DHEA)-preferring, member 1; sulfotransferase hydroxysteroid gene 2; sulfotransferase, DHEA preferring; sulfotransferase, hydroxysteroid preferring 1; sulfotransferase, hydroxysteroid preferring 2; SULT2A1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.