Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SULT2A1 Monoclonal Antibody (OTI4D7), TrueMAB™, OriGene
SDP

Catalog No. 502542092
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2092 30 μL
50-167-2815 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2092 Supplier OriGene Supplier No. TA501694S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SULT2A1 Monoclonal antibody specifically detects SULT2A1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen SULT2A1
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI4D7
Concentration 0.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SULT2A1
Gene Accession No. Q06520
Gene Alias alcohol sulfotransferase; alcohol/hydroxysteroid sulfotransferase; bile salt sulfotransferase; Bile salt sulfotransferase 1; bile-salt sulfotranasferase 2A1; bile-salt sulfotransferase 2A1; Dehydroepiandrosterone sulfotransferase; DHEAS; DHEA-ST; HST; hSTa; Hydroxysteroid sulfotransferase; hydroxysteroid sulfotransferase a; mSTa1; Smp-2; ST; ST2; ST-20; ST2A1; ST2A3; STa; Sta1; Std; Sth1; Sth2; Sulfotransferase 2A1; sulfotransferase family 2A member 1; sulfotransferase family 2A, dehydroepiandrosterone (DHEA)-preferring, member 1; sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone (DHEA) -preferring, member 1; sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone (DHEA)-preferring, member 1; sulfotransferase hydroxysteroid gene 2; sulfotransferase, DHEA preferring; sulfotransferase, hydroxysteroid preferring 1; sulfotransferase, hydroxysteroid preferring 2; SULT2A1
Host Species Mouse
Immunogen Full length human recombinant protein of human SULT2A1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6822
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.