Learn More
SULT2A1 Mouse anti-Human, Clone: OTI7G10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501693

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene is primarily expressed in liver and adrenal tissues where the encoded protein sulfates steroids and bile acids.Specifications
SULT2A1 | |
Monoclonal | |
Unconjugated | |
SULT2A1 | |
DHEA-ST, DHEAS, HST, ST2, ST2A1, ST2A3, STD, hSTa | |
Mouse | |
Affinity Chromatography | |
RUO | |
6822 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI7G10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q06520 | |
SULT2A1 | |
Full length human recombit protein of human SULT2A1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.