Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SULT4A1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981648
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1648 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1648 Supplier Bioss Supplier No. BS11763R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure(number and length of exons) is similar among family members. SULT4A1 isoforms a and b are selectively expressed in brain tissue. Isoform a is the predominant of the two isoforms of this protein. Isoform b includes an alternate segment, compared to isoform a, that causes a frameshift. The resulting protein (isoform b) has a distinct and shorter C-terminus when compared to isoform a.

Specifications

Antigen SULT4A1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SULT4A1
Gene Accession No. Q9BR01
Gene Alias 2400007A17Rik; AI853543; BOS_6206; brain sulfotransferase 4A1; Brain sulfotransferase-like protein; BRSTL1; BR-STL-1; DJ388M5.3; hBR-STL; hBR-STL-1; mBR-STL; nervous system cytosolic sulfotransferase; Nervous system sulfotransferase; NST; rBR-STL; ST4A1; Sulfotransferase 4A1; sulfotransferase family 4A member 1; sulfotransferase family 4A, member 1; sulfotransferase-related protein; SULT4A1; SULTX3; Unknown (protein for MGC:136828); zgc:136828
Gene Symbols SULT4A1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SULT4A1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25830
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.