Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SUMF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558213
Description
Immunogen sequence: MDAYEVSNTE FEKFVNSTGY LTEAEKFGDS FVFEGMLSEQ VKTNIQQAVA AAPWWLPVKG ANWRHPEGPD STILHR Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.
This gene encodes an enzyme that catalyzes the hydrolysis of sulfate esters by oxidizing a cysteine residue in the substrate sulfatase to an active site 3-oxoalanine residue, which is also known as C-alpha-formylglycine. Mutations in this gene cause multiple sulfatase deficiency, a lysosomal storage disorder. Alternative splicing results in multiple transcript variants.
Specifications
SUMF1 | |
Polyclonal | |
Unconjugated | |
SUMF1 | |
AA543204; AAPA3037; AI463102; AI851573; C-alpha-formylglycine-generating enzyme 1; Fge; FGly-generating enzyme; Formylglycine-generating enzyme; MGC131853; MGC150436; OTTHUMP00000115300; PSEC0152; sulfatase modifying factor 1; sulfatase-modifying factor 1; SUMF1; UNQ3037; UNQ3037/PRO9852 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
285362 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NBK3 | |
SUMF1 | |
Recombinant protein corresponding to Human SUMF1. Recombinant protein control fragment (Product #RP-96345). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction