Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUMF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547492
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547492 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547492 Supplier Invitrogen™ Supplier No. PA547492
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human (rh) SUMF2 and less than 5% cross-reactivity with rhSUMF1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

TRUSTED_SUSTAINABILITY

Specifications

Antigen SUMF2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene SUMF2
Gene Accession No. Q8BPG6
Gene Alias 2610040F05Rik; C-alpha-formyglycine-generating enzyme 2; C-alpha-formylglycine-generating enzyme 2; DKFZp566I1024; DKFZp686I1024; DKFZp686L17160; DKFZp781L1035; FGE2; Inactive C-alpha-formylglycine-generating enzyme 2; MGC99485; Paralog of formylglycine-generating enzyme; paralog of the formylglycine-generating enzyme; pFGE; PSEC0171; sulfatase modifying factor 2; sulfatase-modifying factor 2; Sumf2; UNQ1968/PRO4500
Gene Symbols SUMF2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Sulfatase Modifying Factor 2/SUMF2 Gln34-Leu308.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67902
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.