Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUMO-3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520960
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520960 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520960 Supplier Invitrogen™ Supplier No. PA520960
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is mouse liver tissue lysate. PA5-20960 can be used with blocking peptide PEP-1074.

Small ubiquitin-related modifier proteins are members of the ubiquitin-like protein family. SUMO-3 regulates Amyloid beta generation and may be critical in the onset or progression of Alzheimer's disease. SUMO-2/3 forms poly-. SUMO-1, 2, and 3 proteins localize to the nuclear membrane, nuclear bodies, and cytoplasm, respectively. SUMOylation is essential for mammalian cell cycle regulation, SUMO-2/3 localizes to centromeres and condensed chromosomes where binds CENP-E.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUMO-3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SUMO3
Gene Accession No. P55854, Q5XIF4, Q9Z172
Gene Alias 2810014B19Rik; D10Ertd345e; small ubiquitin like modifier 3; small ubiquitin-like modifier 3; small ubiquitin-related modifier 3; SMT3 (supressor of mif two, 3) homolog 1; SMT3 homolog 1; SMT3 suppressor of mif two 3 homolog 1; SMT3 suppressor of mif two 3 homolog 3; SMT3 suppressor of mif two 3 homolog 3 (yeast); Smt3A; Smt3B; SMT3H1; SUMO-2; Sumo3; SUMO-3; Ubiquitin-like protein SMT3A; ubiquitin-like protein SMT3B
Gene Symbols SUMO3
Host Species Rabbit
Immunogen A 13 amino acid peptide near the carboxy terminus of human SUMO3.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20610, 499417, 6612
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.