Learn More
Invitrogen™ SUMO-3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590651
Description
Immunogen sequence: MSEEKPKEGV KTENDHINLK VAGQDGSVVQ FKIKRHTPLS KLMKAYCERQ GLSMRQIRFR FDGQPINETD TPAQLEMEDE DTIDVFQQQT GG; Positive Samples: U-87MG, LO2, Mouse liver, Mouse kidney, Mouse small intestine, Mouse heart, Rat liver, Rat heart; Cellular Location: Cytoplasm, Nucleus, PML body.
Small ubiquitin-related modifier proteins are members of the ubiquitin-like protein family. SUMO-3 regulates Amyloid beta generation and may be critical in the onset or progression of Alzheimer's disease. SUMO-2/3 forms poly-. SUMO-1, 2, and 3 proteins localize to the nuclear membrane, nuclear bodies, and cytoplasm, respectively. SUMOylation is essential for mammalian cell cycle regulation, SUMO-2/3 localizes to centromeres and condensed chromosomes where binds CENP-E.
Specifications
SUMO-3 | |
Polyclonal | |
Unconjugated | |
SUMO3 | |
2810014B19Rik; D10Ertd345e; small ubiquitin like modifier 3; small ubiquitin-like modifier 3; small ubiquitin-related modifier 3; SMT3 (supressor of mif two, 3) homolog 1; SMT3 homolog 1; SMT3 suppressor of mif two 3 homolog 1; SMT3 suppressor of mif two 3 homolog 3; SMT3 suppressor of mif two 3 homolog 3 (yeast); Smt3A; Smt3B; SMT3H1; SUMO-2; Sumo3; SUMO-3; Ubiquitin-like protein SMT3A; ubiquitin-like protein SMT3B | |
Rabbit | |
Affinity chromatography | |
RUO | |
20610, 499417, 6612 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.1 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P55854, Q5XIF4, Q9Z172 | |
SUMO3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-92 of human SUMO3 (NP_0088672). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.