Learn More
Description
Specifically detects SUMO Activating Enzyme E1 (SAE1/UBA2) in Bovine, Canine, Human, Mouse, Rat samples, and it is validated for ELISA, Western Blotting
Specifications
Specifications
| Antigen | SUMO Activating Enzyme E1 (SAE1) |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Concentration | 0.41 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2ug/ml, ELISA 1:5000-1:20000 |
| Formulation | 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide |
| Gene Accession No. | Q9UBE0 |
| Gene Alias | AOS1activator of SUMO1, FLJ3091, HSPC140, Sua1, SUA1sentrin/SUMO-activating protein AOS1, SUMO-1 activating enzyme E1 N subunit, SUMO1 activating enzyme subunit 1, SUMO-1 activating enzyme subunit 1, SUMO-activating enzyme subunit 1, Ubiquitin-like 1-activating enzyme E1A, ubiquitin-like protein SUMO-1 activating enzyme, UBLE1A |
| Gene Symbols | SAE1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
