Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUMO Activating Enzyme E1 (SAE1) Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2442200.05MG
Description
SUMO Activating Enzyme E1 (SAE1) Polyclonal specifically detects SUMO Activating Enzyme E1 (SAE1) in Human samples. It is validated for Western Blot, ELISA.Specifications
SUMO Activating Enzyme E1 (SAE1/UBA2) | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% sodium azide | |
This purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region of human SUMO Activating Enzyme E1 (SAE1/UBA2) Antibody. | |
Primary | |
Human, Mouse, Rat, Bovine, Canine | |
IgG |
Western Blot, ELISA | |
0.41 mg/mL | |
Western Blot 2 μ/mL, ELISA 1:5000-1:20000 | |
Rabbit | |
0.05 mg | |
10055 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction