Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUMO Activating Enzyme E1 (SAE1) Antibody (OTI6B11), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274410AF647
Description
SUMO Activating Enzyme E1 (SAE1) Monoclonal specifically detects SUMO Activating Enzyme E1 (SAE1) in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SUMO Activating Enzyme E1 (SAE1) | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SAE1 | |
Full length recombinant protein of human SUMO Activating Enzyme E1 (SAE1) (NP_005491) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI6B11 | |
Western Blot, Immunohistochemistry | |
AOS1activator of SUMO1, FLJ3091, HSPC140, Sua1, SUA1sentrin/SUMO-activating protein AOS1, SUMO-1 activating enzyme E1 N subunit, SUMO1 activating enzyme subunit 1, SUMO-1 activating enzyme subunit 1, SUMO-activating enzyme subunit 1, Ubiquitin-like 1-activating enzyme E1A, ubiquitin-like protein SUMO-1 activating enzyme, UBLE1A | |
Mouse | |
Affinity Purified | |
Phospho Specific, Stem Cell Markers | |
10055.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction