Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUMO Activating Enzyme E1 (SAE1), p Ser185 Antibody, Novus Biologicals™
SDP

Catalog No. NBP178069 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178069 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP178069 Supplier Novus Biologicals Supplier No. NBP178069
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SUMO Activating Enzyme E1 (SAE1) Polyclonal specifically detects SUMO Activating Enzyme E1 (SAE1) in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen SUMO Activating Enzyme E1 (SAE1) (p Ser185)
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1.02 mg/ml
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, ELISA 1:5000-1:25000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. Q9UBE0
Gene Alias AOS1activator of SUMO1, FLJ3091, HSPC140, Sua1, SUA1sentrin/SUMO-activating protein AOS1, SUMO-1 activating enzyme E1 N subunit, SUMO1 activating enzyme subunit 1, SUMO-1 activating enzyme subunit 1, SUMO-activating enzyme subunit 1, Ubiquitin-like 1-activating enzyme E1A, ubiquitin-like protein SUMO-1 activating enzyme, UBLE1A
Gene Symbols SAE1
Host Species Rabbit
Immunogen This purified SUMO Activating Enzyme E1 (SAE1) [p Ser185] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a region surrounding S185 of the human SUMO Activating Enzyme E1 (SAE1) (SAE1) protein. (Uniprot: Q9UBE0)
Molecular Weight of Antigen 38.45 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10055
Test Specificity This purified antibody is directed against human SUMO Activating Enzyme E1 protein. This antibody is specific for human SAE1 protein phosphorylated at S185. A BLAST analysis using the sequence of the immunizing peptide was used to suggest that this antibody would react with SUMO Activating Enzyme E1 protein from human (100%), bovine, dog, chimpanzee (96%), mouse (93%), and rat (92%) based on a high degree of sequence homology. Cross reactivity against this protein from other sources has not been determined.
Target Species Human, Mouse, Rat, Bovine, Canine
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.