Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUMO Activating Enzyme E1 (SAE1) Rabbit anti-Human, Mouse, Rat, Clone: 0L2N2, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP31531720UL
This item is not returnable.
View return policy
Description
SUMO Activating Enzyme E1 (SAE1) Monoclonal antibody specifically detects SUMO Activating Enzyme E1 (SAE1) in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
SUMO Activating Enzyme E1 (SAE1) | |
Monoclonal | |
Unconjugated | |
AOS1activator of SUMO1, FLJ3091, HSPC140, Sua1, SUA1sentrin/SUMO-activating protein AOS1, SUMO-1 activating enzyme E1 N subunit, SUMO1 activating enzyme subunit 1, SUMO-1 activating enzyme subunit 1, SUMO-activating enzyme subunit 1, Ubiquitin-like 1-activating enzyme E1A, ubiquitin-like protein SUMO-1 activating enzyme, UBLE1A | |
A synthetic peptide corresponding to a sequence within amino acids 200-300 of human SUMO Activating Enzyme E1 (SAE1) (Q9UBE0). | |
20 μg | |
Stem Cell Markers | |
10055 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
0L2N2 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction