Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUMO1 Antibody (5G3), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | SUMO1 |
---|---|
Clone | 5G3 |
Dilution | Flow Cytometry, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000 |
Applications | Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation |
Classification | Monoclonal |
Description
SUMO1 Monoclonal antibody specifically detects SUMO1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, ImmunoprecipitationSpecifications
SUMO1 | |
Flow Cytometry, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
DAP1, GAP modifying protein 1, GAP-modifying protein 1, GMP1SMT3CSMT3H3OFC10UBL1PIC1, SENP2, sentrin, small ubiquitin-related modifier 1, SMT3, SMT3 homolog 3, SMT3 suppressor of mif two 3 homolog 1 (S. cerevisiae), SMT3 suppressor of mif two 3 homolog 1 (yeast), Smt3C, SUMO-1, Ubiquitin-homology domain protein PIC1, ubiquitin-like 1 (sentrin), Ubiquitin-like protein SMT3C, Ubiquitin-like protein UBL1 | |
A synthesized peptide derived from Human SUMO1 [UniProt P63165] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
5G3 | |
Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Alzheimers Research, Core ESC Like Genes, Neuroscience, Stem Cell Markers | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
7341 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title