Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUMO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520300
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520300 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520300 Supplier Invitrogen™ Supplier No. PA520300
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is HL60 cell lysate. PA5-20300 can be used with blocking peptide PEP-0424.

SUMO1 is an ubiquitin-like protein that can be covalently attached to proteins as a monomer or a lysine-linked polymer. Covalent attachment, via an isopeptide bond, to its substrates requires prior activation by the E1 complex SAE1-SAE2 and linkage to the E2 enzyme UBE2I, and can be promoted by E3 ligases such as PIAS1-4, RANBP2 or CBX4. This post-translational modification on lysine residues of proteins plays a crucial role in a number of cellular processes such as nuclear transport, DNA replication and repair, mitosis and signal transduction. SUMO1 is involved, for instance, in targeting RANGAP1 to the nuclear pore complex protein RANBP2. Polymeric SUMO1 chains are also susceptible to polyubiquitination which functions as a signal for proteasomal degradation of modified proteins. SUMO1 may also regulate a network of genes involved in palate development. Mutations in the gene can result in non-syndromic orofacial cleft 10.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUMO1
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SUMO1
Gene Accession No. P63165, P63166, Q5I0H3
Gene Alias CG4494-PA; DAP1; GAP modifying protein 1; GAP-modifying protein 1; GMP1; LD07775p; OFC10; OK/SW-cl.43; OTTHUMP00000202403; OTTHUMP00000202404; OTTHUMP00000202405; OTTHUMP00000202406; OTTHUMP00000202409; OTTHUMP00000202410; PIC1; SENP2; sentrin; Sentrin-1; small ubiquitin-like modifier 1; small ubiquitin-related modifier 1; small ubiquitin-related modifier-1; SMT3; SMT3 homolog 3; SMT3 suppressor of mif two 3 homolog 1; SMT3 suppressor of mif two 3 homolog 1 (yeast); Smt3C; Smt3h3; SMTP3; Sumo1; SUMO-1; SUMO-1 related peptidase; Ubiquitin-homology domain protein PIC1; ubiquitin-like 1; ubiquitin-like 1 (sentrin); ubiquitin-like protein SMT3C; Ubiquitin-like protein UBL1; UBL1
Gene Symbols SUMO1
Host Species Rabbit
Immunogen A 14 amino acid peptide from near the amino terminus of human sumo.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22218, 301442, 7341
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.