Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ SUMO2/3, Rat, Unlabeled, Clone: 3H12,

Catalog No. MABS1139MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABS1139MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1139MI Supplier MilliporeSigma™ Supplier No. MABS1139
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Small ubiquitin-related modifier 2 (UniProt P61956; a.k.a. HSMT3, Sentrin-2, SMT3 homolog 2, Smt3B, SUMO-2, Ubiquitin-like protein SMT3B) and 3 (UniProt P55854; a.k.a. SMT3 homolog 1, Smt3A, SUMO-3, Ubiquitin-like protein SMT3A) are encoded by the SUMO2 (a.k.a. SMT3B, SMT3H2; Gene ID 6613) and SUMO3 (a.k.a. SMT3A, SMT3H1; Gene ID 6612) genes in human. SUMOylation, protein posttranslation modification by small ubiquitin-like modifier (SUMO), is a signaling event in many cellular processes. SUMO proteins are translated as immature precursors and subsequently converted to their mature forms through the activity of sentrin/SUMO-specific proteases (SENPs). As is the case with ubiquitination, SUMOylation is a reversible process. SUMO E1 activating enzyme, E2 conjugating enzyme, and E3 ligase mediate SUMOylation of substrate proteins, while SENPs are responsible for the deSUMOylation. SUMOylation usually occurs at lysine residues in the consensus KxD/E motif, although not all such lysines become SUMOylated and SUMOylation can also occur on lysine residues outside of this motif. SUMO2 and 3 share 97% identity at the amino acid level, while SUMO1 shares approximately 50% and SUMO4 shares about 87% identity with SUMO2/3. SUMO4 is the only SUMO member whose ctivation and conjugation has not been demonstrated. In addition to difference in their target substrates, SUMO2/3 can be SUMOylated and form chains, whereas SUMO1 cannot and may serve as chain terminator.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUMO2/3
Applications Western Blot
Classification Monoclonal
Clone 3H12
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Rat IgG2a hybridoma culture supernatant with 0.05% Sodium Azide.
Gene SUMO2, SMT3B, SMT3H2, SUMO3, SMT3A, SMT3H1
Gene Accession No. P61956;P55854
Host Species Rat
Immunogen GST-tagged recombinant full-length human SUMO3 (Uchimura, Y., et al. (2006). J. Biol. Chem. 281(32):23180-23190).
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_008868;NP_008867
Test Specificity Clone 3H12 detects SUMO2/3 and SUMO2/3-modified (SUMOylated) proteins. Clone 3H12 reacts with SUMO2/3, but not SUMO1 (Uchimura, Y., et al. (2006). J. Biol. Chem. 281(32):23180-23190).
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.