Learn More
Invitrogen™ SUMO2/SUMO3 Polyclonal Antibody

Description
This antibody is predicted to react with bovine, chicken, hamster, porcine, non-human primate, rat and Xenopus and zebrafish based on sequence homology.
Specifications
Specifications
| Antigen | SUMO2/SUMO3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Sumo3 |
| Gene Accession No. | P55854, P61956, P61957, P61959, Q5XIF4, Q9Z172 |
| Gene Alias | 2810014B19Rik; D10Ertd345e; GMP1; HSMT3; I79_000932; MIF2 suppressor; Sentrin; sentrin 2; sentrin-2; small ubiquitin-like modifier 2; small ubiquitin-like modifier 3; small ubiquitin-like modifier 3 L homeolog; small ubiquitin-related modifier 2; Small ubiquitin-related modifier 3; Smt3; SMT3 (supressor of mif two, 3) homolog 1; SMT3 homolog 1; SMT3 homolog 2; SMT3 homolog 3; SMT3 suppressor of mif two 3 homolog 1; SMT3 suppressor of mif two 3 homolog 2; SMT3 suppressor of mif two 3 homolog 2 (yeast); SMT3 suppressor of mif two 3 homolog 3; SMT3 suppressor of mif two 3 homolog 3 (yeast); SMT3 supressor of mif two 3 homolog 2; SMT3A; Smt3B; SMT3C; SMT3H1; SMT3H2; SMT3H3; SUMO-1; SUMO2; SUMO-2; Sumo3; SUMO-3; sumo3.L; Ubiquitin-like protein SMT3A; ubiquitin-like protein SMT3B; UBL1 |
| Gene Symbols | SUMO2, Sumo3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.