Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUMO4 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297570F
Description
SUMO4 Polyclonal specifically detects SUMO4 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
SUMO4 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
dJ281H8.4, IDDM5, small ubiquitin-like modifier 4 protein, Small ubiquitin-like protein 4, small ubiquitin-related modifier 4, SMT3 suppressor of mif two 3 homolog 2, SMT3 suppressor of mif two 3 homolog 4 (S. cerevisiae), SMT3 suppressor of mif two 3 homolog 4 (yeast), SMT3H4, SUMO-4 | |
Produced in rabbits immunized with E. coli-derived Human SUMO4 fragment. | |
0.1 mL | |
Protein Phosphatase | |
387082 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
FITC | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction