Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ SUN 11602
Basic fibroblast growth factor (bFGF) mimetic; neuroprotective
Specifications
Purity | >98% |
---|---|
Product Type | SUN 11602 |
Molecular Weight (g/mol) | 451.61 |
Inhibitors | SUN 11602 |
Description
Basic fibroblast growth factor (bFGF) mimetic. Mimics the neuroprotective effects of bFGF; prevents glutamate-induced neuronal death. Increases levels of CALB1 gene expression in cerebrocortical neurons. Triggers phosphorylation of FGFR, ERK1 and ERK2.Specifications
>98% | |
SUN 11602 | |
451.61 | |
SUN 11602 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title