Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Sun, Earth, and Moon Orbital Model PROMO
SCI_ED

Catalog No. S12895W25
Click to view available options
Height (English):
13 in.
This item is not returnable. View return policy
This item is not returnable. View return policy

Multiple moving axis on this gear driven Sun, Earth, and Moon Orbital Model demonstrate daylight, night, seasons, and phases of moon.

  • Mechanical gear-driven model
  • Multiple moving axis to demonstrate daylight, night, seasons, and phases of the moon
  • 12 V light bulb inside sun demonstrates sunlight on moon and earth
  • Base labeled with the months of the year and seasons for each hemisphere
  • Requires 2 AA batteries, not included

Specifications

Height (Metric) 330 mm
Height (English) 13 in.
Product Type Sun, Earth and Moon Model
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.