Learn More
Thermo Scientific™ SUN-SRi™ Magnetic Screw-Thread Closures
Catalog No. 22030205
Click to view available options
Septum:
With PTFE/Silicone Septa
Sun-SRI provides high quality vials, caps, seals, septa, and closures to meet or exceed original manufacturer's equipment specifications and optimize efficiencies in the laboratory.
  • 18mm closures fit both the 10 and 20mL screw-top headspace vials
  • Can be used with Gerstel™, CTC/Leap™ Technologies, and other magnetic transport autosamplers
TRUSTED_SUSTAINABILITY
Specifications
Material (Liner) PTFE/Silicone
Septum With PTFE/Silicone Septa
Closure Size 18 mm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.