Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SUN-SRi™ SunFlo™ Dissolution Filters

Catalog No. 14823269
Change view
Click to view available options
Porosity:
10 μm
20 μm
35 μm
45 μm
70 μm
Packaging Quantity:
100 Pack
Case of 1000
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Porosity Packaging Quantity
14-823-269 10 μm Case of 1000
14-823-270 70 μm 100 Pack
14-823-274 20 μm Case of 1000
14-823-268 10 μm 100 Pack
14-823-272 45 μm Case of 1000
14-823-267 35 μm Case of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-823-269 Supplier Thermo Scientific™ Supplier No. 400 105
Add to Cart
Add to Cart

Manufactured of Ultra High Molecular Weight (UHMW) foamed porous polyethylene

  • Full flow design removes small particulates
  • Fit securely on the end of a 1/8 in. cannula or sample probe

TRUSTED_SUSTAINABILITY

Specifications

Porosity 10 μm
Packaging Quantity Case of 1000
For Use With 1/8 in. Cannula or Probe
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.