Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUN1 Antibody (12.10F), Alexa Fluor™ 532, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP259943AF532

Catalog No. NP29943A32


Only null left
Add to Cart

Description

Description

SUN1 Monoclonal specifically detects SUN1 in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated.
Specifications

Specifications

SUN1
Monoclonal
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
SUN1
GST fused to residues encoded by exons 6, 8, 10
0.1 mL
Primary
Specificity of mouse, rat SUN1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Store at 4°C in the dark.
Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown
12.10F
Western Blot, Immunocytochemistry/Immunofluorescence
KIAA0810UNC84AFLJ12407, MGC176649, Protein unc-84 homolog A, Sad1 and UNC84 domain containing 1, Sad1 unc-84 domain protein 1, Sad1/unc-84 protein-like 1, SUN domain-containing protein 1, unc-84 homolog A, unc-84 homolog A (C. elegans)
Mouse
Protein A purified
modENCODE Antibodies
23353
Mouse, Rat
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.