Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ SUN2, Mouse, Unlabeled, Clone: 3.1E,

Catalog No. MABT880MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABT880MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT880MI Supplier MilliporeSigma™ Supplier No. MABT880
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SUN domain-containing protein 2 (UniProt Q9UH99; also known as Protein unc-84 homolog B, Rab5-interacting protein, Rab5IP, Sad1/unc-84 protein-like 2) is encoded by the SUN2 (also known as FRIGG, KIAA0668, RAB5IP, UNC84B) gene (Gene ID 25777) in human. It is a single-pass nuclear envelope transmembrane protein that is highly expressed in heart, lung, and muscle. It has a predicted transmembrane domain and a C-terminal region with similarity to the S. pombe spindle pole body protein Sad1. It may also facilitate a nuclear-centrosomal interaction required for nuclear migration and anchorage. It anchors chromosome movement in the prophase of meiosis and is required for telomere attachment to nuclear envelope and gametogenesis. SUN2 protein may also function on endocytic vesicles as a receptor for RAB5-GDP and participate in the activation of RAB5. Slight overexpression of SUN2 protein is seen in heart tissues form patients with congenital heart defects.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUN2
Applications Western Blot
Classification Monoclonal
Clone 3.1E
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG1κ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene SUN2, FRIGG, KIAA0668, RAB5IP, UNC84B
Gene Accession No. Q9UH99
Host Species Mouse
Immunogen GST-tagged recombinant human SUN2 N-terminal LMNA-binding region fragment.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_001186509.1
Test Specificity Target specificity of clone 3.1E was verified by immunocytochemistry and Western blotting analyses using fibroblasts and heart tissue samples from Sun2-knockout mice.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.