Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110745
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110745 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110745 Supplier Invitrogen™ Supplier No. PA5110745
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TDVPQKSRQL YAIIAEYGSR LYKYQARLRM PKEQLELLKK ESQNLENNFR QILFLIEQID VLKALLRDMK DGMDNNHNWN THGDPVEDPD HTEEVSNLVN Highest antigen sequence indentity to the following orthologs: Rat - 69%, Mouse - 72%.

As a probable component of the LINC (LInker of Nucleoskeleton and Cytoskeleton) complex, involved in the connection between the nuclear lamina and the cytoskeleton. The nucleocytoplasmic interactions established by the LINC complex play an important role in the transmission of mechanical forces across the nuclear envelope and in nuclear movement and positioning. May be involved in nuclear remodeling during sperm head formation in spermatogenenis. A probable SUN3:SYNE1 LINC complex may tether spermatid nuclei to posterior cytoskeletal structures such as the manchette.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUN3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SUN3
Gene Accession No. Q8TAQ9
Gene Alias hypothetical protein MGC33329; LOW QUALITY PROTEIN: SUN domain-containing protein 3; Sad1 and UNC84 domain containing 1; Sad1 and UNC84 domain containing 3; sad1/unc-84 domain-containing protein 1; SUN domain-containing protein 3; SUN3; SUNC1
Gene Symbols SUN3
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human SUN3. Recombinant protein control fragment (Product #RP-89513).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 256979
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.