Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570021
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570021 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570021 Supplier Invitrogen™ Supplier No. PA570021
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

As a probable component of the LINC (LInker of Nucleoskeleton and Cytoskeleton) complex, involved in the connection between the nuclear lamina and the cytoskeleton. The nucleocytoplasmic interactions established by the LINC complex play an important role in the transmission of mechanical forces across the nuclear envelope and in nuclear movement and positioning. May be involved in nuclear remodeling during sperm head formation in spermatogenenis. A probable SUN3:SYNE1 LINC complex may tether spermatid nuclei to posterior cytoskeletal structures such as the manchette.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUN3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SUN3
Gene Accession No. Q8TAQ9
Gene Alias hypothetical protein MGC33329; LOW QUALITY PROTEIN: SUN domain-containing protein 3; Sad1 and UNC84 domain containing 1; Sad1 and UNC84 domain containing 3; sad1/unc-84 domain-containing protein 1; SUN domain-containing protein 3; SUN3; SUNC1
Gene Symbols SUN3
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human SUNC1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 256979
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.