Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corporation SunFire Prep C8 OBD Column

Catalog No. 50782145 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
50 mm
75 mm
100 mm
150 mm
250 mm
Diameter (Metric):
19 mm
30 mm
50 mm
Particle Size:
5 μm
10 μm
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Particle Size
50-782-145 250 mm 19 mm 5 μm
50-781-343 100 mm 19 mm 5 μm
50-781-342 50 mm 19 mm 5 μm
50-781-344 150 mm 19 mm 5 μm
50-781-346 50 mm 30 mm 5 μm
50-781-347 75 mm 30 mm 5 μm
50-781-348 100 mm 30 mm 5 μm
50-781-354 150 mm 19 mm 10 μm
50-781-355 250 mm 19 mm 10 μm
50-781-356 150 mm 30 mm 10 μm
50-781-357 250 mm 30 mm 10 μm
50-781-358 150 mm 50 mm 10 μm
50-781-359 250 mm 50 mm 10 μm
50-781-366 150 mm 30 mm 5 μm
50-781-411 50 mm 50 mm 5 μm
50-781-413 100 mm 50 mm 5 μm
50-781-415 50 mm 50 mm 10 μm
50-465-993 50 mm 30 mm 10 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 50-782-145 Supplier Waters Corporation Supplier No. 186004028
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SunFire C8 columns are used for general purpose method development at low pH.

  • The C8 ligand is preferred for some assays because it is less hydrophobic and therefore less retentive compared to SunFire C18.

Specifications

Diameter (Metric) 19 mm
Length (Metric) 250 mm
Max. Pressure 6000 psi (415 Bar)
Particle Shape Spherical
Particle Size 5 μm
Pore Size 100 Å
Quantity 1/Ea.
USP Type L7
Endcapped Yes
Product Type Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.