Learn More
Sigma Aldrich SunTronic™ conductive silver ink for screen printing
Catalog No. 90187925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
Catalog No. Quantity
90-187-925G 25 g
1 options
Catalog No. 90-187-925G Supplier Sigma Aldrich Supplier No. 90187925G
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
SunTronic Conductive Silver Ink AST6025 is a screen printable conductive ink specifically designed for printing fine line and fine pitch conductive traces. The ink exhibits high viscosity, low slumping, excellent conductivity and environmental stability, and adhesion to various substrates, including glass, various metals, PET, and transparent conductive films.AST6025 is specifically suitable for printing of fine interconnects, printed RF antennae, and bus bars for touch panels.SunTronic is a registered trademark of Sun Chemical, Inc.
Specifications
Quantity 25 g
Synonym Ag ink; SunTronic™ AST6025; conductive ink; silver ink
Biological Activity Central nervous system