Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUOX Antibody (OTI5A2), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274411F
Description
SUOX Monoclonal specifically detects SUOX in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
SUOX | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
SUOX | |
Recombinant protein fragment corresponding to amino acids 320-545 of human SUOX (NP_000447) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Immunohistochemistry | |
OTI5A2 | |
Immunohistochemistry | |
EC 1.8.3.1, sulfite oxidase, sulfite oxidase, mitochondrial | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
6821.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction