Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUOX Antibody (SR2065), Novus Biologicals™
SDP

Catalog No. NB259820 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259820 100 μg
NB260077 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB259820 Supplier Novus Biologicals Supplier No. NBP322435100UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SUOX Monoclonal antibody specifically detects SUOX in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen SUOX
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone SR2065
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias EC 1.8.3.1, sulfite oxidase, sulfite oxidase, mitochondrial
Host Species Rabbit
Immunogen A synthesized peptide derived from human SUOX (Uniprot #: P51687)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 6821
Target Species Human, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.