Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUOX Antibody (OTI5A2), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274411AF350
Description
SUOX Monoclonal specifically detects SUOX in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| SUOX | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SUOX | |
| Recombinant protein fragment corresponding to amino acids 320-545 of human SUOX (NP_000447) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI5A2 | |
| Western Blot, Immunohistochemistry | |
| EC 1.8.3.1, sulfite oxidase, sulfite oxidase, mitochondrial | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 6821.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction