Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SUOX Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558294
Description
Immunogen sequence: RLHVVGAPGG QSLSLSLDDL HNFPRYEITV TLQCAGNRRS EMTQVKEVKG LEWRTGAIST ARWAGARLCD VLAQAGHQLC ETEAHVCFEG LDSDPTGT Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.
Sulfite oxidase is a homodimeric protein localized to the intermembrane space of mitochondria. Each subunit contains a heme domain and a molybdopterin-binding domain. The enzyme catalyzes the oxidation of sulfite to sulfate, the final reaction in the oxidative degradation of the sulfur amino acids cysteine and methionine. Sulfite oxidase deficiency results in neurological abnormalities which are often fatal at an early age. Alternative splicing results in multiple transcript variants encoding identical proteins.
Specifications
| SUOX | |
| Polyclonal | |
| Unconjugated | |
| SUOX | |
| SO; sulfite oxidase; sulfite oxidase, mitochondrial; Suox | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6821 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P51687 | |
| SUOX | |
| Recombinant protein corresponding to Human SUOX. Recombinant protein control fragment (Product #RP-102816). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction