Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUOX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558295
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558295 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558295 Supplier Invitrogen™ Supplier No. PA558295
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RDGETVESGE VTIKGYAWSG GGRAVIRVDV SLDGGLTWQV AKLDGEEQRP RKAWAWRLWQ LKAPVPAGQK ELNIVCKAVD DGYNVQPDTV APIWNLRGVL SNAWHRVHVY V Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

Sulfite oxidase is a homodimeric protein localized to the intermembrane space of mitochondria. Each subunit contains a heme domain and a molybdopterin-binding domain. The enzyme catalyzes the oxidation of sulfite to sulfate, the final reaction in the oxidative degradation of the sulfur amino acids cysteine and methionine. Sulfite oxidase deficiency results in neurological abnormalities which are often fatal at an early age. Alternative splicing results in multiple transcript variants encoding identical proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUOX
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SUOX
Gene Accession No. P51687
Gene Alias SO; sulfite oxidase; sulfite oxidase, mitochondrial; Suox
Gene Symbols SUOX
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SUOX. Recombinant protein control fragment (Product #RP-97764).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6821
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.