Learn More
MilliporeSigma™ Supelco™ Supelcarb™ HC Hydrocarbon Trap
The best trap available for removing hydrocarbons and other organics from carrier, fuel, air, and other gas streams
$401.00 - $1018.00
Specifications
Product Line Supelcarb™
Type Hydrocarbon Trap
For Use With (Application) Removing Hydrocarbons and Other Organics From Carrier, Fuel, Air, and Other Gas Streams
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (Metric) Size Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Size Price Quantity & Availability  
11-100-4069
View Documents
MilliporeSigma™ Supelco™
24448
120 cc 1/8 in.
Each for $401.00
Only null left
Add to Cart
11-100-4070
View Documents
MilliporeSigma™ Supelco™
24449
120 cc 1/4 in.
Each for $412.50
Only null left
Add to Cart
11-100-3480
View Documents
MilliporeSigma™ Supelco™
24565
750 cc 1/2 in.
Each for $1,018.00
Only null left
Add to Cart
Description
Description
  • Primarily used for trapping hydrocarbons
  • Used in GC for carrier gas purification
  • Removes organics from carrier gases, air, and hydrogen
  • Adsorbent has twice the trapping ability of activated charcoal
Specifications
Specifications
Supelcarb™
Hydrocarbon Trap
Removing Hydrocarbons and Other Organics From Carrier, Fuel, Air, and Other Gas Streams
Videos
SDS
Documents
Documents
Product Certifications
Supelcarb is a registered trademark of Sigma-Aldrich Co. LLC
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.