Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ SUPELCOSIL™ LC-Diol HPLC Column, 5 μm Particle Size

Catalog No. 111003201 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter (Metric):
4 mm
4.6 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
11-100-3201 4 mm
11-100-3199 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-3201 Supplier MilliporeSigma™ Supelco™ Supplier No. 58201C40
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LC-Diol columns can be used to separate proteins by gel filtration chromatography.

When operated with an aqueous buffer, the diol phase can effectively shield the silica surface from interacting with proteins. A well-known use of diol columns, under normal phase conditions, is the separation of steroids and sterols.
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4 mm
Length (Metric) 25 cm
Particle Size 5 μm
Pore Size 120 Å
Surface Area 170 m2/g
Stationary Phase Diol
Product Line SUPELCOSIL
Matrix Diol Phase/Silica Gel, Spherical Particle Platform
Endcapped No
Phase Reversed Phase
Material Stainless Steel
Technique Normal Phase HPLC, HILIC, SEC
Show More Show Less

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.