Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ SUPELCOSIL™ Suplex™ pKb-100 HPLC Column, 5 μm Particle Size

Catalog No. 111003145 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
15 cm
25 cm
Diameter (Metric):
3 mm
4 mm
4.6 mm
10 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
11-100-3145 25 cm 3 mm
11-100-3130 25 cm 4 mm
11-102-0717 15 cm 4.6 mm
11-102-0709 25 cm 4.6 mm
11-100-2933 25 cm 10 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 11-100-3145 Supplier MilliporeSigma™ Supelco™ Supplier No. 58934C30
Add to Cart
Add to Cart

Suplex™ pKb-100 columns feature the same bonded phase functionality as SUPELCOSIL™ LC-ABZ columns.

Suplex™ pKb-100 columns feature the same bonded phase functionality as SUPELCOSIL™ LC-ABZ columns. These specially deactivated columns differ in that Suplex™ pKb-100 is not endcapped, while SUPELCOSIL™ LC-ABZ is endcapped. The absence of the end-capping reagent results in better performance from Suplex™ pKb-100 for the strongest basic compounds, while LC-ABZ is preferred when the sample also contains acids and/or zwitterions.
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 3 mm
Length (Metric) 25 cm
Particle Size 5 μm
Pore Size 120 Å
Surface Area 170 m2/g
Stationary Phase Amide, Alkyl
Product Line Suplex
Matrix Amide, Alkyl Phase/Silica Gel/Spherical Particle Platform
Endcapped No
Phase Reversed Phase

SUPELCOSIL and Suplex are trademarks of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.