Learn More
MilliporeSigma™ Supelco™ Supelpure™ HC Hydrocarbon Trap
The best trap available for removing hydrocarbons and other organics from carrier, fuel, air, and other gas streams
Supplier: MilliporeSigma™ Supelco™ 22446
Description
- Primarily used for trapping hydrocarbons
- Used in GC for carrier gas purification
- Contains a carbon molecular sieve that also restricts the elution of moisture and oxygen
- Surges of moisture and oxygen that enter the gas line during the installation of a new gas cylinder are released slowly, and diluted by the clean carrier gas from the new cylinder
- Activated charcoal adsorbs hydrocarbons and other contaminants from carrier gases, air, and hydrogen
- Particularly removes C3 and other higher hydrocarbons
- Operates efficiently for approximately six months (when total hydrocarbons in the incoming gas average 10 ppm)
Specifications
120 cc | |
Hydrocarbon Trap | |
Carbon Molecular Sieve | |
11.125 in. | |
1 3/8 in. |
Supelpure™ | |
Removing Hydrocarbons and Other Organics From Carrier, Fuel, Air, and Other Gas Streams | |
1/4 in. | |
28.2 cm | |
35 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Supelpure is a registered trademark of Sigma-Aldrich Co. LLC
Spot an opportunity for improvement?Share a Content Correction